missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-5 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
818.00 SEK - 30750.00 SEK
Specifications
Accession Number | Q8NG35 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 245908, 504180 |
Molecular Weight (g/mol) | 5.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17852073
|
Gibco™
300-68-100UG |
100 μg |
6150.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17812053
|
Gibco™
300-68-1MG |
1 mg |
30750.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872473
|
Gibco™
300-68-20UG |
20 μg |
2015.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17822453
|
Gibco™
300-68-250UG |
250 μg |
10770.00 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17892473
|
Gibco™
300-68-500UG |
500 μg |
18430.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17802483
|
Gibco™
300-68-5UG |
5 μg |
818.00 SEK
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Specifications
Q8NG35 | |
protein with no preservative | |
5.8 | |
E. coli | |
RUO | |
DEFB105A, DEFB105B | |
Unconjugated | |
GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
245908, 504180 | |
Human BD-5 | |
-20°C | |
<1 EU/μg | |
Human | |
Recombinant | |
Lyophilized |