Learn More
Novus Biologicals™ Human Brain Occipital Lobe Whole Tissue Lysate (Adult Whole Normal)
Brand: Novus Biologicals™ NB820-59190
This item is not returnable.
View return policy
Description
The Human Brain Occipital Lobe Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Specifications
Western Blot | |
1 mg | |
Total Protein - Human Adult Normal Tissue: Brain: Occipital Lobe | |
Normal | |
Human | |
Human Brain Occipital Lobe Whole Tissue Lysate (Adult Normal) | |
Adult |
Nervous | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only