missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL5/RANTES Quantikine ELISA Kit
Human CCL5/RANTES Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
7735.00 SEK - 44075.00 SEK
Specifications
Assay Range | 31.20 - 2000pg/mL |
---|---|
Assay Sensitivity | 6.6pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
18782064
|
R&D Systems™
DRN00B |
1 Plate | Sufficient materials to run ELISAs on at least one 96-well plate |
7735.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18762264
|
R&D Systems™
SRN00B |
6 Plates | Sufficient materials to run ELISAs on at least six 96-well plates |
44075.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Human CCL5/RANTES Quantikine ELISA Kit quantifies human CCL5/RANTES in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).Specifications
31.20 - 2000pg/mL | |
HRP | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine | |
Human | |
6352 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
6.6pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Natural and Recombinant Human RANTES | |
ELISA | |
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL),Urine (100uL) | |
chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title