missing translation for 'onlineSavingsMsg'
Learn More

Human Coronavirus Spike Protein Mouse anti-HCoV-OC43, Clone: 15, Novus Biologicals™

Mouse Monoclonal Antibody

5760.00 SEK

Specifications

Antigen Human Coronavirus Spike Protein
Clone 15
Dilution Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18337174
View Documents
Bio-Techne
NBP3-14656-100UL
100 μg
5760.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-OC43 samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Human Coronavirus Spike Protein
Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000
Monoclonal
Purified
RUO
0.2μm filtered solution in PBS without Preservative
3200426
IgG1
Protein A purified
15
Western Blot, ELISA
Unconjugated
Mouse
HCoV-OC43
HKU1, Spike glycoprotein
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.