Learn More
Gibco™ Human FGF-16 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-29-250UG
Additional Details : Weight : 0.16500kg
Description
FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.Specifications
O43320 | |
protein with no preservative | |
23.6 | |
250 μg | |
-20°C | |
<1 EU/μg | |
FGF16 | |
Unconjugated | |
AEVGGVFASL DWDLHGFSSS LGNVPLADSP GFLNERLGQI EGKLQRGSPT DFAHLKGILR RRQLYCRTGF HLEIFPNGTV HGTRHDHSRF GILEFISLAV GLISIRGVDS GLYLGMNERG ELYGSKKLTR ECVFREQFEE NWYNTYASTL YKHSDSERQY YVALNKDGSP REGYRTKRHQ KFTHFLPRPV DPSKLPSMSR DLFHYR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
8823 | |
Human FGF-16 | |
E. coli | |
RUO | |
FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4 | |
Human | |
Recombinant | |
Lyophilized |