Learn More
Applied Biosystems™ Human, Forward Primer, Desalted
Human, Forward Primer, Desalted
Brand: Applied Biosystems™ A15629
This item is not returnable.
View return policy
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.Specifications
Single Tube, Dried-down | |
Forward PCR/SEQ Primer, 25 nmole, Solid, Desalted Purification, Hs00196555_CE Primer ID, 8nmole Minimal Spec Yield, Non-tailed Primer 5'-tail Option, Human Species, Single Tube, Delivered in a Single tube, Dried-down, Used for PCR and Cycle Sequencing Reactions | |
Human | |
Forward PCR/SEQ | |
HPLC | |
Solid |
Mass Spectrometry | |
Forward PCR or SEQ Primer | |
Desalted | |
25 nmol | |
PCR and Cycle Sequencing Reactions |
For Research Use Only. Not for use in diagnostic procedures.