Learn More
Novus Biologicals™ Human Heart Interventricular Septum Whole Tissue Lysate (Adult Whole Hypertension)
Brand:  Novus Biologicals™ NB820-59313
Product Code. 18222865
  • 6700.00 SEK / 1mg
Explore more special offers
Description
Description
The Human Heart Interventricular Septum Whole Tissue Lysate (Adult Whole Hypertension) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - hypertension: Interventricular Septum
Hypertension
Human
Human Heart Interventricular Septum Whole Tissue Lysate (Adult Hypertension)
Adult
Heart
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
For Research Use Only