Learn More
Invitrogen™ Human IL-10 ProQuantum Immunoassay Kit
ProQuantum Immunoassay Kit
Brand: Invitrogen™ A355905
Additional Details : Weight : 2.00000kg
Description
Human IL-10 ProQuantum Immunoassay Kit
ProQuantum Immunoassay Kit
Specifications
Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C. | |
5 x 96 Tests |