Learn More
R&D Systems™ Human LAR (NP_569707) VersaClone cDNA
R&D Systems™ Human LAR (NP_569707) VersaClone cDNA is a member of the protein tyrosine phosphatase (PTP) family.
Brand: R&D Systems™ RDC2115
This item is not returnable.
View return policy
Description
The Human LAR (NP_569707) VersaClone cDNA clone contains the complete ORF for LAR/PTPRF, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LAR/PTPRF insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
LAR/PTPRF | |
10 μg at 0.2 μg/μL | |
5710bp | |
5792 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_569707 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
LCA-homolog, protein tyrosine phosphatase, receptor type, F, PTPRF, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F | |
Human |