Learn More
R&D Systems Human Luminex™ Discovery Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems LXSAHM-30
42084.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
 View more versions of this product
Product Code. 18782214
  • 52605.00 SEK / 1 set
Explore more special offers
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 Plex
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Human
GZMB