missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems Human Luminex™ Discovery Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand: R&D Systems LXSAHM-31
43156.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
31 Plex | |
Multiplex Cytokine Assay Kits | |
Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Human | |
GZMB |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction