Learn More
R&D Systems™ Human Mesenchymal Stem Cell Multi-Color Flow Kit
For flow cytometric verification of human mesenchymal stem/stromal cell identity using four fluorochrome-conjugated antibodies
Brand:  R&D Systems™ FMC002
Additional Details : Weight : 0.09700kg
Product Code. 18175051
  • 9834.52 SEK / 25 tests
Estimated Shipment: 28-08-2024
to see stock.
Description
Description
Variations in techniques as well as differences in the MSC starting population may account for experimental variability and some of the contradictory data in the literature. By clearly defining the starting cell populations, researchers can decrease experimental variability and ultimately decrease apparent discrepancies in the literature that stem from the use of phenotypically different cell populations.
Verifies MSC identity in less than 2 hours
Included conjugated antibodies allow for single step staining
Defines the starting cell population
Multiple markers reduce experimental variation
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.