Learn More
Invitrogen™ PEI-Transferrinfection Kit
Nucleic acid delivery system based on transferrin receptor-mediated endocytosis to carry DNA into cells and the gene transfer efficiency of PEI/DNA complexes
Brand: Invitrogen™ BMS1003
Includes:
- Transferrin-Polyethylenimine-conjugate, lyophilized, 1 mg/mL upon reconstitution; iron incorporated
- Deferrioxamine, lyophilied, 50 mM upon reconstitution
- HBS buffer concentrate (20x)
- (400mM Hepes, pH 7.3, 3M NaCl)
Description
- Method results in a 30 to 1000-fold enhanced transfection efficiency depending on the cell line
- An extremely gentle DNA transfection method which employs physiological uptake mechanisms of the cell
- Transfection efficacy depends on the cell type, the level of surface transferrin receptor expression
- Very high transfection rates have been shown for the tested tumor cell lines B16F10 melanoma, Neuro 2A neuroblastoma and a variety of primary human melanoma cell lines
- In other established cell lines, such as HeLa, CHO, Jurkat, K562, HepG2 and COS the PEI-Transferrinfection works with high efficiences, excellent reproducibility and with the advantage of being an extremely gentle procedure
Specifications
Chemically Defined | |
96-well Plate | |
Transferrin Receptor-mediated Endocytosis | |
Transferrinfection Kit |
Tumor Cell Lines, Established Cell Lines | |
Plasmid DNA | |
Transfection | |
32 Transferrinfections |
User Guide: Polyethylenimine-Transferrinfection (Tf-PEI) Kit
Version: A.0
File size: 131.3 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
363785-002 | Certificate of Analysis | 06/07/2023 | BMS1003 |
363785-001 | Certificate of Analysis | 12/04/2023 | BMS1003 |
337889-005 | Certificate of Analysis | 13/03/2023 | BMS1003 |
337889-004 | Certificate of Analysis | 22/02/2023 | BMS1003 |
337889-003 | Certificate of Analysis | 20/01/2023 | BMS1003 |
Product Content Correction
Product Title
Invitrogen™ PEI-Transferrinfection Kit > 32 transferrinfections
Spot an opportunity for improvement?Share a Content Correction