missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Prolactin Recombinant Protein, PeproTech®
E. coli Recombinant Protein
814.00 SEK - 15320.00 SEK
Specifications
Accession Number | P01236 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 5617 |
Molecular Weight (g/mol) | 23 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17800463
|
Gibco™
100-07-100UG |
100 μg |
3060.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17840363
|
Gibco™
100-07-10UG |
10 μg |
814.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17891633
|
Gibco™
100-07-1MG |
1 mg |
15320.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870363
|
Gibco™
100-07-250UG |
250 μg |
5360.00 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810453
|
Gibco™
100-07-500UG |
500 μg |
9130.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870173
|
Gibco™
100-07-50UG |
50 μg |
2005.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.Specifications
P01236 | |
protein with no preservative | |
23 | |
E. coli | |
RUO | |
AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
5617 | |
Human Prolactin | |
-20°C | |
<1 EU/μg | |
PRL | |
Unconjugated | |
MLPICPGGAA RCQVTLRDLF DRAVVLSHYI HNLSSEMFSE FDKRYTHGRG FITKAINSCH TSSLATPEDK EQAQQMNQKD FLSLIVSILR SWNEPLYHLV TEVRGMQEAP EAILSKAVEI EEQTKRLLEG MELIVSQVHP ETKENEIYPV WSGLPSLQMA DEESRLSAYY NLLHCLRRDS HKIDNYLKLL KCRIIHNNNC | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title