Learn More
Invitrogen™ eBioscience™ Human sCD163 Ready-SET-Go!
Specifically engineered for accurate and precise measurement of human sCD163 protein levels from samples including serum, plasma and cell culture supernatants. CD163 is the 130 kd monocyte/macrophage receptor for haptoglobin–haemoglobin complexes.
Brand: Invitrogen™ 88-50360-22
Includes:
- Capture Antibody Concentrate
- Detection Antibody Concentrate
- Human sCD163 Srandard, lyophilized, 40ng/mL upon reconstitution
- Phosphate Buffered Saline Concentrate (PBS)
- Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Description
Human sCD163 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
0.3125ng/mL | |
Human | |
2 x 96 Tests | |
CD163 |
Cell Culture Supernatant, Plasma, Serum | |
ELISA | |
M130, Hemoglobin Scavenger Receptor, Macrophage marker |
For Research Use Only