Learn More
R&D Systems™ Human SLC16A5/MCT6 (NP_004686.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC3064
Product Code. 18723897
  • 3315.00 SEK / 10µg
Explore more special offers
Specifications
SLC16A5
10 μg at 0.2 μg/μL
1531 bp
MCT 5, MCT 6, MCT5, MCT6, monocarboxylate transporter 5, monocarboxylate transporter 6, solute carrier family 16 (monocarboxylic acid transporters), member 5, solute carrier family 16 member 5, solute carrier family 16, member 5 (monocarboxylic acid transporter 6)
Human
NP_004686.1
cDNA is provided in 10 mM Tris-Cl, pH 8.5
9121
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.