Learn More
R&D Systems™ Human SLC16A5/MCT6 (NP_004686.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC3064
This item is not returnable.
View return policy
Specifications
SLC16A5 | |
10 μg at 0.2 μg/μL | |
1531 bp | |
MCT 5, MCT 6, MCT5, MCT6, monocarboxylate transporter 5, monocarboxylate transporter 6, solute carrier family 16 (monocarboxylic acid transporters), member 5, solute carrier family 16 member 5, solute carrier family 16, member 5 (monocarboxylic acid transporter 6) | |
Human |
NP_004686.1 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
9121 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |