Learn More
Novus Biologicals™ Human Spinal Cord Membrane Tissue Lysate (Adult Membrane Normal)
Brand:  Novus Biologicals™ NB820-60616
Additional Details : Weight : 0.00970kg
Product Code. 18205925
  • 7713.62 SEK / 0.10mg
Estimated Shipment: 14-08-2024
to see stock.
Description
Description
The Human Spinal Cord Membrane Tissue Lysate (Adult Membrane Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
0.1 mg
Membrane Protein - Human Adult Normal Tissue: Spinal Cord
Normal
Human
Human Spinal Cord Membrane Tissue Lysate (Adult Normal)
Adult
Nervous
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Membrane
Human
2 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
Special Offers
For Research Use Only