Learn More
Cytiva HyClone™ Cell Boost™ 5 Supplement
Fed-batch supplement with lipids, amino acids, vitamins, and growth factors for hybridoma, HEK 293, NS0, and CHO cells
Brand: Cytiva SH31117.01
Additional Details : Weight : 0.50000kg
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
- Supports fed-batch cultures with lipids, amino acids, vitamins, and growth factors
- Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line, Human Embryonic Kidney (HEK) 293, NS0, and hybridoma cells
- Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
CHO, HEK293, Hybridoma, NS0 | |
Cell Boost Supplement | |
Improving Productivity and Cell Yield in Fed-Batch Cultures | |
500 mL |
Liquid | |
Bottle | |
HyClone™ |