Learn More
HyClone™ G418 Sulfate
Control or destroy destructive microorganisms with HyClone™ G418 Sulfate.
1405.00 SEK - 4086.12 SEK
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11596471
|
HyClone products Cytiva
SV30069.01 |
G418 Sulfate Solution, 50mg/mL | 20 mL (50 mg/mL) | N/A | |||||
11586471
|
HyClone products Cytiva
SV30068.02 |
G418 Sulfate, >90% Purity | 5 g | N/A | |||||
Description
- Aminoglycoside is used as a selective agent of transfected mammalian, yeast, plant and bacteria cells
- Related to Gentamycin
- Interferes with the function of 80S ribosomes and protein synthesis in eukaryotic cells
- Resistance is conferred by the bacterial gene for aminoglycoside-3'-phosphotransferase that can be expressed in eukaryotic cells
- Common working concentration for G418 is 300 to 1000μg/mL
Specifications
Selection agent |