missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Hydrocortisone
Enhances proliferation and differentiation of endothelial, epithelial, mesenchymal, and oligodendrocyte cells
832.95 SEK - 3296.70 SEK
Specifications
CAS | 125-04-2 |
---|---|
Synonym | Cortisol 21-hemisuccinate; Hydrocortisone sodium succinate; ST51037292; U-4905 |
Molecular Formula | C25H33O8 · Na |
Purity | ≥98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17160332
|
STEMCELL Technologies
74142 |
100 mg |
832.95 SEK
100mg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17170332
|
STEMCELL Technologies
74144 |
1 g |
3296.70 SEK
1g |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Hydrocortisone is a glucocorticoid hormone with anti-inflammatory and immunosuppressive effects (Kang et al; Langhoff & Ladefoged)
- It is also commonly used as a supplement for endothelial, epithelial, mesenchymal, or oligodendrocyte cell culture media, as it supports growth and differentiation
- This product is supplied as the sodium salt of hydrocortisone 21-hemisuccinate, which is a more water-soluble form of hydrocortisone
- Hydrocortisone is required as a supplement in MyeloCult™ H5100 (Human; Catalog #05100) and MyeloCult™ M5300 (Mouse; Catalog #05300), for long-term cultures and the long-term culture-initiating cell (LTC-IC) assay
Specifications
125-04-2 | |
Cortisol 21-hemisuccinate; Hydrocortisone sodium succinate; ST51037292; U-4905 | |
C25H33O8 · Na | |
≥98% |