missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥48%, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Concentration | ≥48% |
Density | 1.15g/mL at 25°C |
Residue after Ignition | ≤0.0005% (as SO4) |
Vapor Pressure | 25mmHg (20°C) |
Quantity | 5 L |
UN Number | 1790 |
IUPAC Name | hydrogen fluoride |
Formula Weight | 20.01g/mol |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Hydrofluoric Acid, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'