missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 500L Liner - Diameter: 31in. (79cm)
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.
Brand: Thermo Scientific™ SH30647.07
This item is not returnable.
View return policy
Description
Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
Features:
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications of the Thermo Scientific tank liner:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
Industry Standard Cylindrical Tanks | |
Non-sterile | |
Fewer resources used and less waste | |
HyPerforma™ | |
Irradiated |
Plastic Film | |
CX3-9 | |
Placed in strong cardboard cartons with product and lot identification. | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |