missing translation for 'onlineSavingsMsg'
Learn More

ICA 069673

KV7.2/KV7.3 channel opener

1520.00 SEK - 5085.00 SEK

Specifications

Chemical Name or Material N-(2-Chloro-5-pyrimidinyl)-3,4-difluorobenzamide
CAS 582323-16-8
Assay Percent Range >99%
Molecular Formula C11H6ClF2N3O
Formula Weight Observed MW: 269.63
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18717046
View Documents
Tocris Bioscience
4305/25
25 mg
5085.00 SEK
25mg
Please to purchase this item. Need a web account? Register with us today!
18795356
View Documents
Tocris Bioscience
4305/5
5 mg
1520.00 SEK
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Selective KV7.2/KV7.3 (KCNQ2/3) channel opener (EC50 = 0.69 μM). Exhibits 20-fold selectivity for KV7.2/KV7.3 over KV7.3/KV7.5, with no measurable activity over a panel of cardiac ion channels. Hyperpolarizes resting membrane potential and inhibits spontaneous action potentials in guinea pig detrusor muscle (DSM) isolated cells. Orally active in animal models of epilepsy.
Specifications

Specifications

N-(2-Chloro-5-pyrimidinyl)-3,4-difluorobenzamide
>99%
Observed MW: 269.63
582323-16-8
C11H6ClF2N3O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ICA 069673

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.