missing translation for 'onlineSavingsMsg'
Learn More

ICAM-1 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen 500-P287-1MG

 View more versions of this product

Product Code. 17825263

  • 18720.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: QTSVSPSKVI LPRGGSVLVT CSTSCDQPKL LGIETPLPKK ELLLPGNNRK VYELSNVQED SQPMCYSNCP DGQSTAKTFL TVYWTPERVE LAPLPSWQPV GKNLTLRCQV EGGAPRANLT VVLLRGEKEL KREPAVGEPA EVTTTVLVRR DHHGANFSCR TELDLRPQGL ELFENTSAPY QLQTFVLPAT PPQLVSPRVL EVDTQGTVVC SLDGLFPVSE AQVHLALGDQ RLNPTVTYGN DSFSAKASVS VTAEDEGTQR LTCAVILGNQ SQETLQTVTI YSFPAPNVIL TKPEVSEGTE VTVKCEAHPR AKVTLNGVPA QPLGPRAQLL LKATPEDNGR SFSCSATLEV AGQLIHKNQT RELRVLYGPR LDERDCPGNW TWPENSQQTP MCQAWGNPLP ELKCLKDGTF PLPIGESVTV TRDLEGTYLC RARSTQGEVT RKVTVNVLSP RYE. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human ICAM-1. Anti-Human ICAM-1-specific antibody was purified by affinity chromatography employing an immobilized Human ICAM-1 matrix. Sandwich ELISA: To detect Human ICAM-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human ICAM-1 (500-P287Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human ICAM-1. Western Blot: To detect Human ICAM-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human ICAM-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICAM-1
Polyclonal
Unconjugated
ICAM1
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells
Rabbit
Antigen affinity chromatography
RUO
3383
-20°C
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P05362
ICAM1
CHO cells-derived Recombinant Human ICAM-1
1 mg
Primary
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers