missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Mouse anti-Human, Clone: 3A5A5, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Human IgA |
---|---|
Clone | 3A5A5 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827075
|
Proteintech
60099-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16817075
|
Proteintech
60099-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes the Chain C region of human Ig alpha-1 (IGHA1).
Ig alpha is the major immunoglobulin class in body secretions. The protein may serve both to defend against local infection and to prevent access of foreign antigens to the general immunologic system.Specifications
Human IgA | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IgA1, IGHA1 | |
IGHA1 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
3A5A5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P01876 | |
3493 | |
Human IgA Fusion Protein Ag7417 | |
Primary | |
-20°C | |
IGHA1 |