missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535343
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and porcine based on sequence homology.
IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.Specifications
IGFBP2 | |
Polyclonal | |
Unconjugated | |
IGFBP2 | |
AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3485 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P18065 | |
IGFBP2 | |
KLH conjugated synthetic peptide between 277-305 amino acids from the C-terminal region of human IGFBP2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |