Learn More
IgG(H+L) Goat anti-Human, FITC, Secondary Antibody, Proteintech
Goat anti-Human Affinipure Secondary Antibody
Brand:  Proteintech SA00003-12-500UL
Product Code. 17680327
  • 1171.54 SEK / 500µL
Explore more special offers
This item is not returnable. View return policy
Specifications
IgG(H+L)
FITC
Goat
500 μL
Human
Liquid
Immunofluorescence, Flow Cytometry
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.