missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IGH/BCL10 DY Translocation FISH Probe
Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.
20380.00 SEK - 33560.00 SEK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | immunoglobulin heavy locus |
Gene Alias | IGH,IGHDY1,MGC72071,MGC88774 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003585
|
Abnova™
FT0025.100uL |
100 μL |
20380.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013585
|
Abnova™
FT0025.200uL |
200 μL |
33560.00 SEK
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
IGH,IGHDY1,MGC72071,MGC88774 | |
3492;8915 | |
Liquid |
Tube | |
immunoglobulin heavy locus | |
IGH | |
DNA/RNA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title