Learn More
Abnova™ IGH/CCND1 DY Translocation FISH Probe
Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FT0020.200uL
 View more versions of this product
Product Code. 16013575
  • 33560.00 SEK / 200µL
Explore more special offers
This item is not returnable. View return policy
Specifications
Store at 4°C in the dark
Fluorescent In Situ Hybridization (Cell)
IGH,IGHDY1,MGC72071,MGC88774
3492;595
200 μL
Tube
immunoglobulin heavy locus
IGH
DNA/RNA