Learn More
Abnova™ IGH/FOXP1 DY Translocation FISH Probe
Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FT0015.200uL
 View more versions of this product
Product Code. 16013565
  • 33560.00 SEK / 200µL
Explore more special offers
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
This item is not returnable. View return policy
Specifications
Store at 4°C in the dark.
Tube
3492/27086
200 μL
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe