missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF2 Mouse anti-Human, Mouse, Clone: 1E6A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66333-1-IG-20UL
This item is not returnable.
View return policy
Description
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Specifications
IGSF2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A8E0Y8, Q93033 | |
Cd101 | |
IGSF2 Fusion Protein Ag23297 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
1E6A7 | |
Unconjugated | |
CD101 | |
CD101, Cell surface glycoprotein V7, EWI 101, EWI101, IGSF2, V7 | |
Mouse | |
Protein G | |
RUO | |
630146, 9398 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IGSF2 Mouse anti-Human, Mouse, Clone: 1E6A7, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction