missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IKK gamma Inhibitor Peptide Set
Applications: In vitro assay, In vivo assay, Functional (Inhibition), Block/Neutralize
Brand: Novus Biologicals™ NBP2-26504

This item is not returnable.
View return policy
Specifications
Human, Mouse, Rat | |
Functional (Inhibition), In vitro assay, In vivo assay | |
2 mg |
1.IKK-gamma NEMO Binding Domain (NBD) Inhibitor Peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKTALDWSWLQTE (IKK-gamma/NEMO binding sequence is underlined). Molecular weight: 3692 2.Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 |
|
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ IKK gamma Inhibitor Peptide Set
Spot an opportunity for improvement?Share a Content Correction