missing translation for 'onlineSavingsMsg'
Learn More

IL-36 gamma/IL-1F9 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1935.00 SEK - 4730.00 SEK

Specifications

Antigen IL-36 gamma/IL-1F9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18323624
View Documents
Bio-Techne
NBP3-03323-20UL
20 μL
1935.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18348316
View Documents
Bio-Techne
NBP3-03323-100UL
100 μL
4730.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-36 gamma/IL-1F9 Polyclonal specifically detects IL-36 gamma/IL-1F9 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IL-36 gamma/IL-1F9
Polyclonal
Purified
RUO
Mouse
IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of mouse IL36G IL-36 gamma/IL-1F9 (NP_705731.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Biologically Active Proteins
PBS with 50% glycerol, pH7.3.
56300
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.