missing translation for 'onlineSavingsMsg'
Learn More

IL-5 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

2205.00 SEK - 19580.00 SEK

Specifications

Antigen IL-5
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17865433
View Documents
Invitrogen
500-P55-50UG
50 μg
2205.00 SEK
50µg
Please to purchase this item. Need a web account? Register with us today!
17845433
View Documents
Invitrogen
500-P55-100UG
100 μg
2830.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
17875643
View Documents
Invitrogen
500-P55-1MG
1 mg
19580.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: MEIPMSTVVK ETLTQLSAHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEMLFQNL SLIKKYIDRQ KEKCGEERRR TRQFLDYLQE FLGVMSTEWA MEG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-5. Anti-Murine IL-5-specific antibody was purified by affinity chromatography employing an immobilized Murine IL-5 matrix. Sandwich ELISA: To detect Murine IL-5 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine IL-5 (500-P55Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-5. Western Blot: To detect Murine IL-5 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine IL-5 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
Specifications

Specifications

IL-5
ELISA, Western Blot
Unconjugated
Rabbit
Mouse
P04401
16191
E.coli-derived Recombinant Murine IL-5
Antigen affinity chromatography
Il5
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Il5
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IL-5 Polyclonal Antibody, PeproTech®, Invitrogen™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.