missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5 Rat anti-Human/Mouse, PE, Clone: TRFK5, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554395
This item is not returnable.
View return policy
Description
The TRFK5 antibody reacts with mouse interleukin-5 (IL-5) and cross-reacts with human IL-5. The TRFK5 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Host Species: Rat
Clone: TRFK5
Isotype: IgG1 κ
Species Reactivity: Mouse, Human
Immunogen: Mouse Semi-Purified T-Cell Clone Supernatant
Intracellular Staining
Specifications
IL-5 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Semi-Purified T-Cell Clone Supernatant | |
0.1 mg | |
Immunology | |
Human, Mouse | |
IgG1 κ |
Flow Cytometry | |
TRFK5 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction