Learn More
IL-7 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P27-100UG
Description
AA Sequence of recombinant protein: MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-7. Anti-Human IL-7-specific antibody was purified by affinity chromatography employing an immobilized Human IL-7 matrix. Sandwich ELISA: To detect Human IL-7 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-7 (500-P27Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-7. Western Blot: To detect Human IL-7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human IL-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications
IL-7 | |
Polyclonal | |
Unconjugated | |
Il7 | |
A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3574 | |
-20°C |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
P13232 | |
Il7 | |
E.coli-derived Recombinant Human IL-7 | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.