Learn More
IL-7R alpha/CD127 Mouse anti-Rat, Clone: 717519, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB8484-100
Encompass
Product Code. 15954051
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Specifications
IL-7R alpha/CD127
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127 Glu21-Asp239 Accession # NP_001099888
100 μg
3575
Reconstitute at 0.5mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
CyTOF, Flow Cytometry
717519
Flow Cytometry 0.25μg/106 cells, CyTOF-ready
NP_001099888
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects rat IL-7 R alpha/CD127in direct ELISAs.
Rat
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
IL-7R alpha/CD127 Mouse anti-Rat, Clone: 717519, R&D Systems™