Learn More
Invitrogen™ IL-8 Human Matched Antibody Pair
Matched Antibody Pair
Brand:  Invitrogen™ BMS204-3MST
Product Code. 15511557
  • 7330.00 SEK / Pack of 10
Explore more special offers
Includes:
  • 1 vial (5.5mL) monoclonal Coating Antibody to human IL-8/NAP-1 (100μg/mL)
  • 1 vial (55μL) Biotin-Conjugate anti-human IL-8/NAP-1 polyclonalantibody
  • 1 vial (11μL) Streptavidin-HRP
  • 1 vial (30μL) human IL-8/NAP-1 Standard protein, 200ng/mL
  • 2 vials (50mL) Sample Diluent
This item is not returnable. View return policy
Description
Description
The Human IL-8/NAP-1 Matched Antibody Pair Kit comes with pre-matched antibody pairs,standards,and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-8. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-8 (Interleukin-8) is a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8,secreted by endothelial cells and monocytes,which acts as a major chemoattractant for neutrophils and also T cells and basophils. IL-8,first isolated from osteosarcoma cells,contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1),granulocyte chemotactic protein 1 (GCP-1),monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-8
Monoclonal/Polyclonal
P10145
CXCL8
RUO
3576
-20°C
ELISA
Biotin
Protein 3-10C, CXCL8, NAP-1, GCP-1
10 x 96 Tests
Primary
Human
ELISA Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ IL-8 Human Matched Antibody Pair > 10 x 96 Tests