Learn More
Zeiss™ Illumination Adapter
Includes light guide (D) for HXP 120 and X-Cite 120
Brand:  Zeiss™ 423302-0000-000
Additional Details : Weight : 1.00000kg
Product Code. 13468938
  • 4610.05 SEK / Each
Estimated Shipment: 17-10-2024
to see stock.
Add to basket
Description
Description
Accessory for Zeiss Microscopes
Specifications
Specifications
Illumination Adapter
HXP 120 and X-Cite 120
1 Ea.
For light sources with light guide
With Light Guide