missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-b]pyridazine, 98%, Thermo Scientific™

Product Code. 15305038
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 766-55-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.12
InChI Key VTVRXITWWZGKHV-UHFFFAOYSA-N
Synonym imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
PubChem CID 136599
IUPAC Name imidazo[1,2-b]pyridazine
SMILES C1=CC2=NC=CN2N=C1

Specifications

Melting Point 54.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material Imidazo[1,2-b]pyridazine
compliance-icons
Product Identifier
  • Imidazo¢1,2-b!pyridazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'