missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15470878
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water. 

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58767-51-4
Molecular Formula C3H3ClN2O2S
Molecular Weight (g/mol) 166.579
MDL Number MFCD08687938
InChI Key MXCZCMCHRWGSBX-UHFFFAOYSA-N
Synonym 1h-imidazole-4-sulfonyl chloride, 4-chlorosulfonylimidazole, imidazole-4-sulfonyl chloride, pubchem19106, imidazole-4-sulfonylchloride, imidazole-4-sulphonyl chloride, imidazole-4-sul-fonyl chloride, 1-h-imidazole-4-sulphonyl chloride
PubChem CID 18961164
IUPAC Name 1H-imidazole-5-sulfonyl chloride
SMILES C1=C(NC=N1)S(=O)(=O)Cl

Specifications

Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 165.96
Percent Purity 97%
Chemical Name or Material Imidazole-4-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.