Learn More
INHBB & ACVR1C Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand:  Abnova DI0042.1Set
Product Code. 16072438
  • 8740.00 SEK / 1 set
Explore more special offers
Description
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the INHBB protein, and the other against the ACVR1C protein for use in in situ Proximity Ligation Assay.
Specifications
Specifications
INHBB
Unconjugated
MGC157939
1 Set
Primary
Antibody pair set content: 1. INHBB rabbit purified polyclonal antibody (20 ug) 2. ACVR1C mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
In situ PLA
inhibin, beta B
INHBB
RUO
3625/130399
Human
Ab Pair
Product Title
INHBB & ACVR1C Protein Protein Interaction Antibody Pair, Abnova™ > 1 Set; Unconjugated