Learn More
INHBB & ACVR1C Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DI0042.1Set
This item is not returnable.
View return policy
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the INHBB protein, and the other against the ACVR1C protein for use in in situ Proximity Ligation Assay.Specifications
INHBB | |
Unconjugated | |
MGC157939 | |
1 Set | |
Primary | |
Antibody pair set content: 1. INHBB rabbit purified polyclonal antibody (20 ug) 2. ACVR1C mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
In situ PLA | |
inhibin, beta B | |
INHBB | |
RUO | |
3625/130399 | |
Human | |
Ab Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
INHBB & ACVR1C Protein Protein Interaction Antibody Pair, Abnova™ > 1 Set; Unconjugated
Spot an opportunity for improvement?Share a Content Correction