Learn More
Thermo Scientific™ Injectors for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Discover our wide range of reliable, screw-in, self-aligning injectors, with differing internal diameters and materials, to match your sample matrices and ICP-MS analysis needs.
8192.31 SEK
Specifications
Type Injector
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17862836
View Documents
Thermo Scientific™
1305600
Injector 2.5 mm ID quartz
8192.31 SEK
Each
Description
Description
Choose from this range of reliable, screw-in, self-aligning injectors, with differing internal diameters and materials, to match your sample matrices and analytical needs.
The iCAP MX and iCAP Q/Qnova Series ICP-MS systems come as standard with a 2.5 mm internal diameter, quartz injector. All of our injectors are easily removable for improved robustness and reduced maintenance requirements.
Choose high-quality consumables to improve performance
  • Quartz injectors are suitable for most aqueous acidic and organic applications
  • Sapphire injectors are suitable for highly concentrated acids
  • Platinum injectors are suitable for ultra-trace and highly concentrated acids
  • Injectors with integrated ports are suitable for dry aerosol introduction, e.g., laser ablation
Specifications
Specifications
Injector