Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand:  Hanna Instruments™ HI95762-03
 View more versions of this product
Product Code. 11657100
  • 1375.00 SEK / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
  • Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
Specifications
Free chlorine ultra low range determination
300 individual packets of HI95762-0
1 Set
Standard Method 4500-Cl G
Compatible Hanna benchtop and portable photometers
300
0.000 to 0.500mg/L free chlorine
Water Testing Kit