Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand: Hanna Instruments™ HI9371103
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
- Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
Total Chlorine determination | |
Individual Packets | |
1 Set | |
USEPA method 330.5 and Standard Method 4500-Cl G |
Compatible Hanna benchtop and portable photometers | |
300 | |
0.00 to 5.00mg/L total chlorine | |
Water Testing Kit |
Safety and Handling
- Total Chlorine Reagents (300 tests)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 1
Hazard Statement
- H332-Harmful if inhaled.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : potassium iodide, EDTA
Your input is important to us. Please complete this form to provide feedback related to the content on this product.