Learn More
Lauda™ Insulation Tube
Manufactured from silicone. Lauda™ Insulation Tube is suitable for hoses with max outer diameter.
Brand:  Lauda™ RKJ 013
Additional Details : Weight : 0.50000kg
Product Code. 10786333
  • 370.00 SEK / Each
Estimated Shipment: 22-11-2024
to see stock.
Description
Description
  • For hoses with max outer diameter
Specifications
Specifications
105°C
EPDM Rubber
29 mm
Insulation Tubing
105°C