Learn More
Lauda™ Insulation Tubing
Brand:  Lauda™ RKJ 023
Additional Details : Weight : 27.00000kg
Product Code. 10306751
  • 505.00 SEK / 1m
Estimated Shipment: 12-08-2024
to see stock.
Description
Description
Order Info
DiameterMetric
Specifications
Specifications
Insulation pipes for insulation posteriori
150°C
Insulation Tubing
19 mm