missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Rabbit anti-Human, Clone: INSR/2277R, Novus Biologicals™
Rabbit Monoclonal Antibody
2815.00 SEK
Specifications
Antigen | Insulin R/CD220 |
---|---|
Clone | INSR/2277R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Insulin R/CD220 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
10 mM PBS with 0.05% BSA | |
3643 | |
IgG | |
Protein A or G purified |
INSR/2277R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
Recombinant fragment of extracellular domain of human Insulin R/CD220 (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title