missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulysin/IDE Rabbit anti-Human, Mouse, Rat, Clone: 8O8F3, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | Insulysin/IDE |
---|---|
Clone | 8O8F3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18399406
|
Bio-Techne
NBP3-15352-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18337144
|
Bio-Techne
NBP3-15352-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulysin/IDE Monoclonal antibody specifically detects Insulysin/IDE in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Insulysin/IDE | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3416 | |
IgG | |
Affinity purified |
8O8F3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Insulysin/IDE (P14735). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title