missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 7, Rabbit anti-Human,Mouse, Clone: FIB27, Novus Biologicals™

Rabbit Monoclonal Antibody

2470.00 SEK - 5400.00 SEK

Specifications

Antigen Integrin beta 7
Clone FIB27
Concentration 1 mg/mL
Dilution Flow Cytometry, Immunoprecipitation, Block/Neutralize
Applications Flow Cytometry, Immunoprecipitation, Blocking Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18064516
View Documents
Bio-Techne
NBP2-81087-0.025MG
0.025 mg
2470.00 SEK
25µg
Please to purchase this item. Need a web account? Register with us today!
18323400
View Documents
Bio-Techne
NBP2-81087-0.2MG
0.2 mg
5400.00 SEK
0.20mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize.
Specifications

Specifications

Integrin beta 7
1 mg/mL
Flow Cytometry, Immunoprecipitation, Blocking Assay
Unconjugated
Rabbit
Signal Transduction
PBS
3695
FIB27 was prepared by immunizing rats with TK1 cells.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
FIB27
Flow Cytometry, Immunoprecipitation, Block/Neutralize
Monoclonal
Purified
RUO
Human, Mouse
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
ITGB7
IgG κ
Protein A purified
FIB27 binds specifically to both human and mouse Integrin beta 7 at epitope region Dl on which is involved in all a4b7-mediated adhesion events. Does not compete with DAKT32 for binding. FIB27 blocks LS722-induced TK1 cell aggregation (mAb LS722 is an activating anti-a4B7 antibody and induces aggregation via an a4B7-dependent pathway). Integrin beta 7 is one subunit is a heterodimer which makes up an intergin molecule. In the mouse beta-7 is selectively found on the majority of mature lymphocytes, whereas a small subpopulation of thymocytes and bone marrow cells express beta-7. Beta-7 is also expressed in variable amounts in CD4 T memory cells. Integrin molecules mediate cell-cell and cell-extracellular matrix adhesion and are involved in lymphocyte homing, leukocyte recruitment to inflammatory sites, myogenesis, hemopoiesis, and melanoma metastasis.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.